Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flufenamic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620758 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-758 5 g
18-620-759 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-758 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F30205GM
Add to Cart
Add to Cart

Synonym: Fluphenamic Acid, Nichisedan, Arlef, Flufacid

Chemical Identifiers

CAS 530-78-9
Molecular Formula C14H10F3NO2
Molecular Weight (g/mol) 281.23
InChI Key LPEPZBJOKDYZAD-UHFFFAOYSA-N
IUPAC Name 2-{[3-(trifluoromethyl)phenyl]amino}benzoic acid
SMILES OC(=O)C1=CC=CC=C1NC1=CC=CC(=C1)C(F)(F)F

Specifications

Quantity 5 g
Percent Purity 97%
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.