Learn More
Hardy Diagnostics Fluid A, USP, 300 mL Fill, Polycarbonate Bottle with Needle Port Septum
Shop All Hardy Diagnostics Products

Description
- For use by the pharmaceutical and medical device industry to ensure sterility of pharmaceuticals and medical devices
- Hardy Diagnostics sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center
- Each lot is certified for pH, sterility and growth promotion using ATCC™ strains specified by the USP
- Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results
Specifications
Specifications
| Form | Liquid |
| Manufacturing Origin | US with Globally Sourced Materials |
| Product Type | Liquid Medium, Peptone |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria |
| Test Type | Media Testing, Microbial Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.