Learn More
MOLTOX Molecular Toxicology Inc Fluid A + 0.5% Tween 80 (PEP w/0.5% Tween 80), pH 7.1 + 0.2, 90 mL/bottle, Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 26659
Encompass
This item has a minimum order quantity of 96 per supplier requirements. Fluid D is peptone water (Fluid A) with the addition of Polysorbate (Tween) 80. It is used for diluting or rinsing when performing sterility testing. Tween 80 acts as a surfactant which will break down any residue present. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. NC0687061