Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics Fluid D, USP, 1000 mL Fill, Polycarbonate Bottle
SDP

Catalog No. 22026374 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Target Organism Class:
Bacteria
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Organism Class
22-026-374 Bacteria
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-026-374 Supplier Hardy Diagnostics Supplier No. U115
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A rinsing and diluting fluid for use in USP testing protocols.

  • Contains peptic digest of animal tissue and polysorbate 80 with a pH of 7.1
  • Features and Benefits:
    • Heavy-duty: Bottles are tough and sturdy to reduce breakage
    • Optically clear: bottles are made of clarified polycarbonate for clear viewing
    • Easy to read: Each bottle is clearly marked with a measuring fill line
    • Accuracy and efficient: Each lot is certified for pH, sterility and growth promotion using ATCC™ strains specified by the USP

Specifications

Form Liquid
Manufacturing Origin US with Globally Sourced Materials
Product Type Liquid Medium, Peptone
Sterility Non-sterile
Target Organism Class Bacteria
Test Type Media Testing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.