Learn More
Electron Microscopy Sciences Fluid Dispenser Bottles, Volume: 180 ml., 1/EA
SDP
Catalog No. 502090873
Encompass_Preferred
These ESD safe polyethylene solvent dispensers are available in three standard sizes. These dispensers have neither a stopper nor a cap to remove. Just touch the dispensing dish with a brush cloth or applicator and solvent is brought to the surface instantly as needed. Stainless steel valves seal contents in bottle until used. The fluid pumps into the dish and it does not drip back into the bottle, keeping stored liquids uncontaminated. Surface resistivity of 10E7 to 10E9 Volume: 180 ml.
Catalog No. 50-209-0873 Supplier Electron Microscopy Sciences Supplier No. 6429907
These ESD safe polyethylene solvent dispensers are available in three standard sizes. These dispensers have neither a stopper nor a cap to remove. Just touch the dispensing dish with a brush cloth or applicator and solvent is brought to the surface instantly as needed. Stainless steel valves seal contents in bottle until used. The fluid pumps into the dish and it does not drip back into the bottle, keeping stored liquids uncontaminated. Surface resistivity of 10E7 to 10E9 Volume: 180 ml.