Learn More
Thermo Scientific™ Fluid Filter Kit
Catalog No. 13100773
Change view
Click to view available options
Type:
Replacement Filter Bags
For Use With (Equipment):
ThermoFlex 7500 or 24000
Partial Flow:
100 micron
250 micron
35 micron
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment) Partial Flow
13-100-773 Replacement Filter Bags ThermoFlex 7500 or 24000 35 micron
13-265-253 Replacement Filter Bags ThermoFlex 7500 or 24000 100 micron
13-100-769 Replacement Filter Bags ThermoFlex 7500 or 24000 250 micron
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-100-773 Supplier Thermo Scientific™ Supplier No. 611000000101
Only null left
Add to Cart
Add to Cart
Fluid Filter Kits are available with 5 to 40μm partial-flow filtration.
Thermo Scientific™ Fluid Filter Kits are available with 5 to 40μm partial-flow filtration.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoFlex 7500 or 24000
Type Replacement Filter Bags
Partial Flow 35 micron
Quantity 2 Ea.