Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ Full Back SMS Isolation Gowns

Catalog No. 19160257 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-160-257 Universal
19-160-258 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-160-257 Supplier Cardinal Health™ Supplier No. 2100PG
Add to Cart
Add to Cart
This item is not returnable. View return policy

SMS lightweight material gowns provide flexible coverage in minimal fluid settings.

Constructed of high-quality, fluid-resistant, tri-layer spunbond meltblown spunbond (SMS) material.

  • Latex-free elastic wrists
  • Waist belt
  • Adjustable ties at neck

Specifications

For Use With (Application) Healthcare, Nursing
Gender Unisex
No. of Pockets None
Packaging Type Flat Pack
Size Universal
Product Type Isolation Gown
Material SMS
Color Yellow
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Model 2100PG
Sleeve Style Long Sleeve
Autoclavable Not Autoclavable
No. per Pack 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.