Learn More
Instech 20ga single channel Plastic Swivel (pink), sterile
SDP
Catalog No. 5080965
Encompass
Plastic Swivels
For Tethered Infusion or Sampling with Mice and Rats
Plastic Swivels from Instech deliver reliable, cost‑effective performance designed for researchers who prefer fresh fluid‑path components, these swivels feature durable plastic bodies paired with the same proven self‑adjusting seals used in our stainless‑steel models—ensuring smooth, leak‑free operation through even long‑term studies.
Each swivel includes a convenient clamp for quick connection to standard Instech spring tethers, with color‑coded bodies for easy gauge identification. Available in three sizes, with or without luer inlets, and supplied EtO‑sterilized or integrated into ready‑to‑use infusion kits. Compatible with PS95 rat teathers.
Features
  • One channel for infusion or sampling
  • Self-adjusting seals
  • Includes plastic swivel-to-tether clamp (20ga mate with PS95-based rat tethers)
  • Individually packaged and EtO sterilized
Catalog No. 50-809-65 Supplier Instech Supplier No. 375/20PS
Plastic Swivels
For Tethered Infusion or Sampling with Mice and Rats
Plastic Swivels from Instech deliver reliable, cost‑effective performance designed for researchers who prefer fresh fluid‑path components, these swivels feature durable plastic bodies paired with the same proven self‑adjusting seals used in our stainless‑steel models—ensuring smooth, leak‑free operation through even long‑term studies.
Each swivel includes a convenient clamp for quick connection to standard Instech spring tethers, with color‑coded bodies for easy gauge identification. Available in three sizes, with or without luer inlets, and supplied EtO‑sterilized or integrated into ready‑to‑use infusion kits. Compatible with PS95 rat teathers.
Features
  • One channel for infusion or sampling
  • Self-adjusting seals
  • Includes plastic swivel-to-tether clamp (20ga mate with PS95-based rat tethers)
  • Individually packaged and EtO sterilized