Learn More
Instech 25ga single channel Stainless Steel Swivel
SDP
Catalog No. 5080969
Encompass
Stainless Steel Swivels
For Tethered Infusion or Sampling with Mice and Rats
This Instech 25ga single channel stainless steel swivels is autoclavable and may be reused hundreds of times. Self-adjusting seals, at the heart of every Instech swivel, actually improve with use. All stainless steel models include a universal swivel-to-tether clamp which mates with a range of Instech spring tethers.
Available in three sizes - 25ga, 22ga and 20ga. The 25ga model has the lowest torque and is recommended for mice, though it is also good for rat studies where low dead volume is important. 22ga is the standard for rats.
Features
  • One channel for tethered infusion or sampling
  • Stainless steel body - autoclavable
  • Available in three sizes - 25ga, 22ga and 20ga
  • Self-adjusting seals
  • Adjustable swivel-to-tether clamp
Catalog No. 50-809-69 Supplier Instech Supplier No. 375/25
Stainless Steel Swivels
For Tethered Infusion or Sampling with Mice and Rats
This Instech 25ga single channel stainless steel swivels is autoclavable and may be reused hundreds of times. Self-adjusting seals, at the heart of every Instech swivel, actually improve with use. All stainless steel models include a universal swivel-to-tether clamp which mates with a range of Instech spring tethers.
Available in three sizes - 25ga, 22ga and 20ga. The 25ga model has the lowest torque and is recommended for mice, though it is also good for rat studies where low dead volume is important. 22ga is the standard for rats.
Features
  • One channel for tethered infusion or sampling
  • Stainless steel body - autoclavable
  • Available in three sizes - 25ga, 22ga and 20ga
  • Self-adjusting seals
  • Adjustable swivel-to-tether clamp