Learn More
Instech 20ga dual channel Stainless Steel Swivel
SDP
Catalog No. 5080971
Encompass
Two Channel Stainless Steel Swivels
For Infusion and Blood Sampling in Rats and Larger Animals
The 375/D/20 is a larger‑ID, two‑channel swivel ideal for blood pressure measurement or IV feeding.
Its dual channels allow simultaneous infusion and withdrawal, independent dual‑line infusion, or infusion on one channel while monitoring on the other. Like the stainless single channel swivels, these swivels have been built to last a lifetime (however, users should follow the instructions carefully when cleaning the swivels after every use).
Each swivel includes a universal clamp compatible with button, harness, and head block tethers. Repair and replacement parts are available.
Features
  • Dual channel
  • Stainless steel body
  • Self-adjusting seals
  • Includes universal swivel-to-tether clamp
Catalog No. 50-809-71 Supplier Instech Supplier No. 375/D/20
Two Channel Stainless Steel Swivels
For Infusion and Blood Sampling in Rats and Larger Animals
The 375/D/20 is a larger‑ID, two‑channel swivel ideal for blood pressure measurement or IV feeding.
Its dual channels allow simultaneous infusion and withdrawal, independent dual‑line infusion, or infusion on one channel while monitoring on the other. Like the stainless single channel swivels, these swivels have been built to last a lifetime (however, users should follow the instructions carefully when cleaning the swivels after every use).
Each swivel includes a universal clamp compatible with button, harness, and head block tethers. Repair and replacement parts are available.
Features
  • Dual channel
  • Stainless steel body
  • Self-adjusting seals
  • Includes universal swivel-to-tether clamp