Learn More
Instech Low-torque dual channel swivel, 22ga OD, 25ga equivalent ID
SDP
Catalog No. 5080973
Encompass
Two Channel Stainless Steel Swivels
For Infusion and Blood Sampling with Mice
375/D/22LT - Low-torque two channel swivel for mice, 22ga.
Its dual channels allow simultaneous infusion and withdrawal, independent dual‑line infusion, or infusion on one channel while monitoring on the other. Like the stainless single channel swivels, these swivels have been built to last a lifetime (however, users should follow the instructions carefully when cleaning the swivels after every use).
Each swivel includes a universal clamp compatible with button, harness, and head block tethers. Repair and replacement parts are available.
Features
  • Dual channel
  • Stainless steel body
  • Self-adjusting seals
  • Includes universal swivel-to-tether clamp
Catalog No. 50-809-73 Supplier Instech Supplier No. 375/D/22LT
Two Channel Stainless Steel Swivels
For Infusion and Blood Sampling with Mice
375/D/22LT - Low-torque two channel swivel for mice, 22ga.
Its dual channels allow simultaneous infusion and withdrawal, independent dual‑line infusion, or infusion on one channel while monitoring on the other. Like the stainless single channel swivels, these swivels have been built to last a lifetime (however, users should follow the instructions carefully when cleaning the swivels after every use).
Each swivel includes a universal clamp compatible with button, harness, and head block tethers. Repair and replacement parts are available.
Features
  • Dual channel
  • Stainless steel body
  • Self-adjusting seals
  • Includes universal swivel-to-tether clamp