Learn More
MilliporeSigma Fluid Thioglycollate in bottle with screw cap and septum, double packed, 12/pk
Catalog No. NC0550116
Encompass
The Fluid Thioglycollate medium is used for sterility testing. It is primarily intended for the detection of anaerobic bacteria. The fluid is double packed for increasing the test method reliability.
Catalog No. NC0550116 Supplier MilliporeSigma Supplier No. STBMFTM12DP
The Fluid Thioglycollate medium is used for sterility testing. It is primarily intended for the detection of anaerobic bacteria. The fluid is double packed for increasing the test method reliability.