Learn More
MOLTOX Molecular Toxicology Inc Fluid Thioglycolate Medium, 200 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948777
Encompass_Preferred
Fluid Thioglycollate Medium (FTM) contains a small amount of agar and several reducing agents which detects the presence of anaerobic bacteria in sterility testing. This medium is recommended by the FDA and in the USP for use in sterility confirmation. Strict aerobes tend to grow at the surface whereas obligate anaerobes grow below the upper oxidized layer portion of the broth. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-777 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26606
Fluid Thioglycollate Medium (FTM) contains a small amount of agar and several reducing agents which detects the presence of anaerobic bacteria in sterility testing. This medium is recommended by the FDA and in the USP for use in sterility confirmation. Strict aerobes tend to grow at the surface whereas obligate anaerobes grow below the upper oxidized layer portion of the broth. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.