Learn More
NEOGEN Fluid Thioglycolate Medium, 5Kg
SDP
Catalog No. 502016003
Encompass_Preferred
Fluid Thioglycolate Medium is used for sterility testing. This medium conforms to Harmonized USP/EP/JP requirements and is not intended for use in the diagnosis of disease or other conditions in humans.Fluid Thioglycolate Medium is recommended by the Harmonized Pharmacopoeia for sterility testing and conforms to USP/EP/JP performance specification. In this formulation casein and yeast extract provide a source of nitrogen, essential vitamins and amino acids. The glucose provides a carbon source and sodium chloride maintains osmotic balance. L-Cystine and sodium Thioglycolate act as reducing agents to create an anaerobic environment and maintain a low Eh. This is aided by the low level of agar which reduces the oxygen permeability through the medium. Resazurin is an oxidation indicator which turns from colorless to red/pink when oxidised.For more information visit https://www.neogen.com/solutions/fluid-thioglycollate-medium-ncm0108
Catalog No. 50-201-6003 Supplier NEOGEN Supplier No. 700004499
Fluid Thioglycolate Medium is used for sterility testing. This medium conforms to Harmonized USP/EP/JP requirements and is not intended for use in the diagnosis of disease or other conditions in humans.Fluid Thioglycolate Medium is recommended by the Harmonized Pharmacopoeia for sterility testing and conforms to USP/EP/JP performance specification. In this formulation casein and yeast extract provide a source of nitrogen, essential vitamins and amino acids. The glucose provides a carbon source and sodium chloride maintains osmotic balance. L-Cystine and sodium Thioglycolate act as reducing agents to create an anaerobic environment and maintain a low Eh. This is aided by the low level of agar which reduces the oxygen permeability through the medium. Resazurin is an oxidation indicator which turns from colorless to red/pink when oxidised.For more information visit https://www.neogen.com/solutions/fluid-thioglycollate-medium-ncm0108