Learn More
Teknova Fluid Thioglycollate Medium Agar Plates. 100mm, 20 Plates per Sleeve, Sterile 4 pack.
SDP
Catalog No. 502387514
Encompass
0.50% Yeast Extract, 1.50% Tryptone, 0.55% Dextrose, 0.05% L-Cystine, 0.25% Sodium Chloride, 0.05% Sodium Thioglycollate, 0.0001% Resazurin,1.50% Agar
Catalog No. 50-238-7514 Supplier Teknova Supplier No. T25004PK
0.50% Yeast Extract, 1.50% Tryptone, 0.55% Dextrose, 0.05% L-Cystine, 0.25% Sodium Chloride, 0.05% Sodium Thioglycollate, 0.0001% Resazurin,1.50% Agar