Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Fluid Thioglycollate Medium (FTM), ETO

Catalog No. B257544 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Fluid thioglycollate medium; Fill volume: 100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B257544 Fluid thioglycollate medium; Fill volume: 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B257544 Supplier BD Supplier No. 257544
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for enrichment and cultivation of anaerobes, aerobes and microaerophiles

  • Useful for microbial load testing of environmentally-controlled areas, clean rooms, pharmaceuticals
TRUSTED_SUSTAINABILITY

Specifications

Appearance Clear
Broth Type Fluid Thioglycollate Medium
Cap Type Screw Cap
Certifications/Compliance EP/USP
Detects Microaerophiles
Form Prepared Medium
Format Bottle
Media Type Fluid
Product Type Culture Medium
Sterility Sterile
Test Type Sterility Testing
Fill Volume 100 mL
Formulation L-Cystine, Sodium Chloride, Dextrose, Yeast Extract, Tryptone, Nitrogen in headspace
For Use With (Application) Cultivation of Anaerobes, Aerobes, and Microaerophiles
Includes Syrup Bottle, Screw Cap w/ Septum and Hole
Packaging Type Syrup Bottle
Quantity 100 mL
Specimen Type Biologics, Food, Water
Manufacturing Quality FDA-registered, DIN/EN/ISO 9001:2000, ISO 13485:2003, cGMP21CFR820
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.