Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium
GSA_VA

Catalog No. R064690 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
16mm x 125mm Tube
16mm x 125mm Tube, Hungate Cap
20mm x 113mm Tube
Borosilicate Bottle - Septum
Bottle
SB
Tube
WMB Lug
Media Type:
Fluid Thioglycollate Medium
Manufacturing Origin:
United States
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R064690 100/Pk. Tube Fluid Thioglycollate Medium United States
R07172 100/Pk. 16mm x 125mm Tube Fluid Thioglycollate Medium United States
R08846 100/Pk. 20mm x 113mm Tube Fluid Thioglycollate Medium United States
R112640 10/Pk. WMB Lug Fluid Thioglycollate Medium United States
R112641 10/Pk. SB Fluid Thioglycollate Medium United States
R112642 10/Pk. Borosilicate Bottle - Septum Fluid Thioglycollate Medium United States
R112646 10/Pk. Bottle Fluid Thioglycollate Medium United States
R117800 20/Pk. 16mm x 125mm Tube, Hungate Cap Fluid Thioglycollate Medium United States
R117801 100/Pk. 16mm x 125mm Tube, Hungate Cap Fluid Thioglycollate Medium United States
R064692 20/Pk. Tube Fluid Thioglycollate Medium United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. R064690 Supplier Thermo Scientific™ Supplier No. R064690
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.

Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes and bottles of Fluid Thioglycollate Medium w/Indicator, Dextrose.

Fluid Thioglycollate Medium is available as dehydrated culture medium or in ready-to-use prepared media tubes and bottles and in formats that meet international industrial guidelines for microbiological testing, including United States Pharmacopeia (USP) guidelines for sterility testing.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, COMPF
Color Clear broth w/visible agar light to medium amber
Content And Storage 2 to 25°C
Format Tube
Manufacturing Origin United States
Media Type Fluid Thioglycollate Medium
Product Type Microbiology Media
Fill Volume 9 mL
Includes w/ Indicator, Dextrose Medium
Packaging Type Glass Tube
Quantity 100/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.