Learn More
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium
GSA_VA
Catalog No. R07172 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
16mm x 125mm Tube
16mm x 125mm Tube, Hungate Cap
20mm x 113mm Tube
Borosilicate Bottle - Septum
Bottle
SB
Tube
WMB Lug
Media Type:
Fluid Thioglycollate Medium
Manufacturing Origin:
United States
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R07172 100/Pk. 16mm x 125mm Tube Fluid Thioglycollate Medium United States
R064690 100/Pk. Tube Fluid Thioglycollate Medium United States
R08846 100/Pk. 20mm x 113mm Tube Fluid Thioglycollate Medium United States
R112640 10/Pk. WMB Lug Fluid Thioglycollate Medium United States
R112641 10/Pk. SB Fluid Thioglycollate Medium United States
R112642 10/Pk. Borosilicate Bottle - Septum Fluid Thioglycollate Medium United States
R112646 10/Pk. Bottle Fluid Thioglycollate Medium United States
R117800 20/Pk. 16mm x 125mm Tube, Hungate Cap Fluid Thioglycollate Medium United States
R117801 100/Pk. 16mm x 125mm Tube, Hungate Cap Fluid Thioglycollate Medium United States
R064692 20/Pk. Tube Fluid Thioglycollate Medium United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. R07172 Supplier Thermo Scientific™ Supplier No. R07172
Add to Cart
Add to Cart
This item is not returnable. View return policy
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.
  • For sterility testing according to USP (United States Pharmacopeia)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, USP.
Format 16mm x 125mm Tube
Manufacturing Origin United States
Media Type Fluid Thioglycollate Medium
Product Type Microbiology Media
Quantity 100/Pk.