Learn More
HIMEDIA Fluid Thioglycollate Medium, M009, Microbiology Culture Media, Dehydrated Culture Media, Fluid Medium, Sterility Testing, Anaerobe Cultivation, Powder, 5 kg
Catalog No. NC4013660
Encompass_Preferred
Fluid Thioglycollate Medium is the standard broth for sterility testing of biologicals and for cultivating anaerobes, aerobes and microaerophiles from pharmaceutical and clinical samples. Sodium thioglycollate and L-cystine lower the redox potential to support anaerobes, while a small amount of agar limits oxygen diffusion and resazurin signals the aerobic zone as a pink band. Tryptone, yeast extract, dextrose and sodium chloride provide rich nutrition, final pH 7.1. It is widely used in USP and EP sterility test workflows. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013660 Supplier HIMEDIA Supplier No. M0095KG
Fluid Thioglycollate Medium is the standard broth for sterility testing of biologicals and for cultivating anaerobes, aerobes and microaerophiles from pharmaceutical and clinical samples. Sodium thioglycollate and L-cystine lower the redox potential to support anaerobes, while a small amount of agar limits oxygen diffusion and resazurin signals the aerobic zone as a pink band. Tryptone, yeast extract, dextrose and sodium chloride provide rich nutrition, final pH 7.1. It is widely used in USP and EP sterility test workflows. Dehydrated powder, store in a cool dry place. Packaged as 500 g.