Learn More
Alkali Scientific Fluid Thioglycollate Medium w/Indole 100mL Bottle, 1/Pk
SDP
Catalog No. 5000662772
Encompass_Preferred
Fluid Thioglycollate Medium is a versatile liquid culture medium used for sterility testing and cultivation of aerobic and anaerobic microorganisms. The formulation creates an oxygen gradient within the tube, allowing different organisms to grow at their preferred oxygen levels. Reducing agents remove dissolved oxygen, while an indicator helps visualize oxygen presence. Aerobic organisms grow near the surface, while anaerobes grow deeper in the medium. This design allows detection of a wide range of microorganisms in a single system. The medium is widely used in pharmaceutical testing, clinical microbiology, and laboratory research for monitoring microbial contamination and growth.
Catalog No. 50-006-62772 Supplier Alkali Scientific Supplier No. FTFB00
Fluid Thioglycollate Medium is a versatile liquid culture medium used for sterility testing and cultivation of aerobic and anaerobic microorganisms. The formulation creates an oxygen gradient within the tube, allowing different organisms to grow at their preferred oxygen levels. Reducing agents remove dissolved oxygen, while an indicator helps visualize oxygen presence. Aerobic organisms grow near the surface, while anaerobes grow deeper in the medium. This design allows detection of a wide range of microorganisms in a single system. The medium is widely used in pharmaceutical testing, clinical microbiology, and laboratory research for monitoring microbial contamination and growth.