Learn More
Alkali Scientific Fluid Thioglycollate Medium w/o Indole w/Hemin & Vitamin K 5mL, 10/Pk
SDP
Catalog No. 5000662867
Encompass_Preferred
Fluid Thioglycollate Medium enriched with hemin and vitamin K is designed for the cultivation of fastidious anaerobic bacteria. These added growth factors support organisms that cannot synthe essential compounds required for metabolism. The medium lacks certain indicators that may interfere with sensitive organisms, improving recovery rates of clinically important anaerobes. It is commonly used in clinical microbiology for isolating bacteria from infections where oxygen sensitive organisms are present. The enriched formulation ensures reliable growth of demanding species and supports accurate diagnosis and laboratory testing.
Catalog No. 50-006-62867 Supplier Alkali Scientific Supplier No. FTFT00
Fluid Thioglycollate Medium enriched with hemin and vitamin K is designed for the cultivation of fastidious anaerobic bacteria. These added growth factors support organisms that cannot synthe essential compounds required for metabolism. The medium lacks certain indicators that may interfere with sensitive organisms, improving recovery rates of clinically important anaerobes. It is commonly used in clinical microbiology for isolating bacteria from infections where oxygen sensitive organisms are present. The enriched formulation ensures reliable growth of demanding species and supports accurate diagnosis and laboratory testing.