Learn More
Thermo Scientific™ FluidEase™ Pro ClipTip™ Electronic Pipette Charging Stand and Extension Kit

Description
The Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic pipette charging stand enables simultaneous wireless charging of up to four electronic pipettes without individual cables. Dedicated resting positions allow users to temporarily place pipettes with tips attached and liquid inside during short workflow interruptions. A modular extension design supports scalable charging capacity for shared laboratory environments.
Features:
- Electronic Pipette Charging Stand for Shared Laboratories: Designed specifically for Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic pipettes used in single- and multichannel workflows.
- Simultaneous Charging of Up to Four Pipettes: Supports wireless charging of up to four electronic pipettes at the same time, reducing cable clutter and bench complexity.
- Dedicated Charging and Resting Positions: Includes active charging positions and separate temporary resting positions for safely placing pipettes during short workflow pauses.
- Temporary Resting with Tips and Liquid Inside: Allows users to briefly rest pipettes with tips attached and liquid retained when reaching for tubes, reagents, or plates.
- Modular and Scalable Design: Extension kits enable charging capacity to scale from one to four pipettes as laboratory needs grow.
- Organized Bench Footprint: Keeps electronic pipettes visible, accessible, and securely positioned to support efficient shared-lab operation.
Specifications
Specifications
| Color | White |
| For Use With | FluidEase Pipette Charging Stand |
| Type | Charging Stand Extension Kit |
| Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.