Learn More
Thermo Scientific™ FluidEase™ Pro Electronic Pipette Wall Charging Stand
Catalog No. 02687124 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With:
FluidEase Pro Pipettes
Catalog No. For Use With
02-687-124 FluidEase Pro Pipettes
1 options
Catalog No. 02-687-124 Supplier Thermo Scientific™ Supplier No. 9420540
Add to Cart
Add to Cart
Thermo Scientific™ FluidEase™ Pro Electronic Pipette Wall Charging Stand provides convenient charging and secure storage while freeing valuable bench space, helping keep pipettes powered, organized, and easily accessible for efficient laboratory workflows.
Make the most of your laboratory space with the Thermo Scientific™ FluidEase™ Pro Electronic Pipette Wall Charging Stand. Multiple wall attachment options provide installation flexibility for different laboratory environments, while integrated charging keeps electronic pipettes ready for use. A dedicated off-bench location supports an organized workspace and convenient pipette access between workflows.
  • Space-Saving Wall Storage: Keep your FluidEase™ Pro electronic pipette conveniently stored and charged off the bench, helping maintain a clear, organized workspace.
  • Four Installation Options: Choose from four included mounting options—magnets, suction cups, adhesive tape, or screws with wall plugs—to position the stand where it works best for your laboratory space and workflow.
  • Integrated Charging: Charge your electronic pipette while it is stored, helping ensure it stays powered and ready for your next workflow.
  • FluidEase™ Pro Pipette Compatibility: One wall charging stand supports all FluidEase™ Pro electronic pipettes, providing a convenient charging solution across your laboratory.
  • Installation Accessories Included: Multiple mounting accessories are included with the stand, giving you the flexibility to select the installation method that best suits your laboratory environment.
  • Straightforward Setup: An easy-to-follow installation guide and essential charging components are included to support quick and straightforward setup.
TRUSTED_SUSTAINABILITY
Specifications
For Use With FluidEase Pro Pipettes
Type Charging Stand