Learn More
Waters Corp Fluidics Selector Valve Rebuild Kit, MS System, As Required Replacement
Catalog No. 502391740 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit is specifically designed for a fluidics selector valve. It is an essential component for the maintenance and repair of various mass spectrometers. The kit ensures the continued optimal performance of your analytical instruments by providing the necessary parts to rebuild the selector valve.
  • Type: Rebuild kit
  • System type: MS (Mass Spectrometer)
  • Replacement schedule: As required
  • Ensures continued optimal performance of analytical instruments
  • Compatible with Synapt G2, Synapt G2-S, Synapt G2-Si, Xevo QTof, Xevo G2, Xevo G2-S, Xevo G2-XS, Vion, Xevo TQ, Xevo TQ-S, and Xevo TQ-XS mass spectrometers.
Catalog No. 50-239-1740 Supplier Waters Corp Supplier No. 700004787
This rebuild kit is specifically designed for a fluidics selector valve. It is an essential component for the maintenance and repair of various mass spectrometers. The kit ensures the continued optimal performance of your analytical instruments by providing the necessary parts to rebuild the selector valve.
  • Type: Rebuild kit
  • System type: MS (Mass Spectrometer)
  • Replacement schedule: As required
  • Ensures continued optimal performance of analytical instruments
  • Compatible with Synapt G2, Synapt G2-S, Synapt G2-Si, Xevo QTof, Xevo G2, Xevo G2-S, Xevo G2-XS, Vion, Xevo TQ, Xevo TQ-S, and Xevo TQ-XS mass spectrometers.