Learn More
Waters Corp Fluidics Tubing and Fitting Kit, Spare Parts Kit, for Mass Spectrometer Systems
Catalog No. 502391028 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides replacement tubing and fittings for the fluidics of a Mass Spectrometer. It is included in the start-up kits for various Mass Spect Spectrometers, including Xevo G2-S, Xevo G2-XS, Synapt G2-S, Synapt G2-Si, Synapt XS, and SELECT SERIES Cyclic IMS.
  • Spare parts kit
  • Designed for mass spectrometer systems
  • Replacement as required
Catalog No. 50-239-1028 Supplier Waters Corp Supplier No. 700008827
This kit provides replacement tubing and fittings for the fluidics of a Mass Spectrometer. It is included in the start-up kits for various Mass Spect Spectrometers, including Xevo G2-S, Xevo G2-XS, Synapt G2-S, Synapt G2-Si, Synapt XS, and SELECT SERIES Cyclic IMS.
  • Spare parts kit
  • Designed for mass spectrometer systems
  • Replacement as required