Learn More
Cole-Parmer Techne F932P Fluidized Sand Bath System, Temperature Range 50-500°C, 2 x 750W Heaters; 120 VAC
Catalog No. NC1526359 Shop All Cole-Parmer Products
Encompass_Preferred
Use these fluidized sand baths for your heat treating processes, temperature sensor calibrations, and electrical component testing. Baths use aluminum oxide fluidized by low-pressure air as a dry bath medium with excellent heat transfer properties. Inert, nonflammable aluminum oxide is safer to use than oil and will not corrode your sample vessels or produce toxic fumes. Vessels of all shapes can be easily immersed and safely removed from the bath, without the hazard and mess of hot oils. Make temperature adjustments via a control knob and reference dial. Baths require a dry air flow of approximately 3 cu ft/minute for model 01184-00 and -02 and 5 cu ft/minute for 01184-04 at about 3 psi pressure; order an air pump separately. All baths have a thermowell and an overspill flange. Note: not recommended for shape setting in laboratory research device manufacturing or for heating of reactors.
Catalog No. NC1526359 Supplier Cole-Parmer Supplier No. 0118400
Add to Cart
Add to Cart
Request a Quote
Use these fluidized sand baths for your heat treating processes, temperature sensor calibrations, and electrical component testing. Baths use aluminum oxide fluidized by low-pressure air as a dry bath medium with excellent heat transfer properties. Inert, nonflammable aluminum oxide is safer to use than oil and will not corrode your sample vessels or produce toxic fumes. Vessels of all shapes can be easily immersed and safely removed from the bath, without the hazard and mess of hot oils. Make temperature adjustments via a control knob and reference dial. Baths require a dry air flow of approximately 3 cu ft/minute for model 01184-00 and -02 and 5 cu ft/minute for 01184-04 at about 3 psi pressure; order an air pump separately. All baths have a thermowell and an overspill flange. Note: not recommended for shape setting in laboratory research device manufacturing or for heating of reactors.