Learn More
Solstice QuEChERS, Alternative SPE Cleanup
Catalog No. 6003122 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Description:
250 mg GCB, 500 mg PSA
500 mg GCB, 500 mg PSA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
60-031-22 250 mg GCB, 500 mg PSA
60-031-23 500 mg GCB, 500 mg PSA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-031-22 Supplier Solstice Supplier No. 5700
6 mL SPE Cleanup Column
  • Efficient method of sample preparation
  • Array of tube sizes and sorbent materials available
  • Appropriate for use with acetonitrile and other high purity solvents
TRUSTED_SUSTAINABILITY
Specifications
Description 250 mg GCB, 500 mg PSA
For Use With (Application) Solid Phase Extraction Cleanup
Volume (Metric) 6 mL
Quantity 30