Learn More
Solstice QuEChERS, Sample Extraction
Catalog No. 6002972 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Sorbent:
4 gm MgSO4, 1 gm sodium chloride per Tube
4 gm MgSO4, 1 gm sodium chloride, 500 mg sodium citrate (dibasic sesquihydrate),1 gm sodium citrate (tribasic dihydrate) per Tube
6 gm MgSO4, 1.5 gm sodium acetate per Tube
Quantity:
50
250
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sorbent Quantity
600-29-72 6 gm MgSO4, 1.5 gm sodium acetate per Tube 250
600-29-66 4 gm MgSO4, 1 gm sodium chloride per Tube 50
600-29-68 4 gm MgSO4, 1 gm sodium chloride per Tube 250
600-29-69 4 gm MgSO4, 1 gm sodium chloride, 500 mg sodium citrate (dibasic sesquihydrate),1 gm sodium citrate (tribasic dihydrate) per Tube 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 600-29-72 Supplier Solstice Supplier No. 5626
Extraction Salt for QuEChERS Method with 50 mL Tube
QuEChERS
  • Quick, Easy, Cheap, Effective, Rugged and Safe Sample Extraction and Cleanup Products
  • Solstice QuEChERS products offer an alternative sample preparation technique to traditional liquid-liquid and solid phase extraction (SPE) methods
  • the use of QuEChERS has the potential to lower the overall cost of sample preparation due to the increased efficiency and ease-of use when compared with SPE methods
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) AOAC QuEChERS Method
Sorbent 6 gm MgSO4, 1.5 gm sodium acetate per Tube
Volume (Metric) 50 mL
Quantity 250