Learn More
INTERWORLD HIGHWAY LLC Fluke 51-2 Thermometer Series II
SDP
Catalog No. NC2883572
Encompass_Preferred
SINGLE INPUT THERMOMETER, 60HZ NOISE REJECTIONRelative time clock on MIN, MAX, and AVG provides a time reference for major eventsElectronic Offset function allows compensation of thermocouple errors to maximize overall accuracyMeasures J, K, T, and E-types of thermocouplesReadout in °C, °F, or Kelvin (K)Splash and dust resistant case protected by impact absorbing holsterUser-friendly front panel is easy to set up and operateSleep mode increases battery life; typical 1000-hour battery lifeBattery door allows easy battery replacement without breaking the calibration sealOptional ToolPak accessory allows the thermometer to hang from any metal object (with the rare earth magnet) or secure around a pipe (with hook-and-loop straps) for hands-free operation3 year warranty
Catalog No. NC2883572 Supplier INTERWORLD HIGHWAY LLC Supplier No. 674686
SINGLE INPUT THERMOMETER, 60HZ NOISE REJECTIONRelative time clock on MIN, MAX, and AVG provides a time reference for major eventsElectronic Offset function allows compensation of thermocouple errors to maximize overall accuracyMeasures J, K, T, and E-types of thermocouplesReadout in °C, °F, or Kelvin (K)Splash and dust resistant case protected by impact absorbing holsterUser-friendly front panel is easy to set up and operateSleep mode increases battery life; typical 1000-hour battery lifeBattery door allows easy battery replacement without breaking the calibration sealOptional ToolPak accessory allows the thermometer to hang from any metal object (with the rare earth magnet) or secure around a pipe (with hook-and-loop straps) for hands-free operation3 year warranty