Learn More
Interworld Highway LLC Tequipment Net FLUKE 52-2 THERMOMETER SER II
SDP
Catalog No. NC2242884
Encompass_Preferred
Large backlit dual display shows any combination of T1, T2 (52 and 54 only), T1-T2 (52 and 54 only), plus MIN, MAX, or AVGRelative time clock on MIN, MAX, and AVG provides a time reference for major eventsElectronic Offset function allows compensation of thermocouple errors to maximize overall accuracyMeasures J, K, T, and E-types of thermocouplesReadout in °C, °F, or Kelvin (K)Splash and dust resistant case protected by impact absorbing holsterUser-friendly front panel is easy to set up and operateSleep mode increases battery life; typical 1000-hour battery lifeBattery door allows easy battery replacement without breaking the calibration sealOptional ToolPak accessory allows the thermometer to hang from any metal object (with the rare earth magnet) or secure around a pipe (with hook-and-loop straps) for hands-free operation3 year warranty
This item is not returnable. View return policy
Catalog No. NC2242884 Supplier Interworld Highway LLC Tequipment Net Supplier No. 674689
Large backlit dual display shows any combination of T1, T2 (52 and 54 only), T1-T2 (52 and 54 only), plus MIN, MAX, or AVGRelative time clock on MIN, MAX, and AVG provides a time reference for major eventsElectronic Offset function allows compensation of thermocouple errors to maximize overall accuracyMeasures J, K, T, and E-types of thermocouplesReadout in °C, °F, or Kelvin (K)Splash and dust resistant case protected by impact absorbing holsterUser-friendly front panel is easy to set up and operateSleep mode increases battery life; typical 1000-hour battery lifeBattery door allows easy battery replacement without breaking the calibration sealOptional ToolPak accessory allows the thermometer to hang from any metal object (with the rare earth magnet) or secure around a pipe (with hook-and-loop straps) for hands-free operation3 year warranty