Learn More
ZORO TOOLS FLUKE-561 INFRARED THERMOMETER
Catalog No. NC2000584
Encompass
Infrared Thermometer, Backlit LCD, -40 Degrees to 1022 Degrees F, Single Dot Laser Sighting
Catalog No. NC2000584 Supplier ZORO TOOLS Supplier No. G3013622
Infrared Thermometer, Backlit LCD, -40 Degrees to 1022 Degrees F, Single Dot Laser Sighting