Learn More
INTERWORLD HIGHWAY LLC Fluke 80PK-1 Bead Probe
SDP
Supplier:  INTERWORLD HIGHWAY LLC 750422
Encompass_Preferred
PROBE, THERMOCOUPLE, BEADED K-TYPEType-K thermocouple for general purpose applicationsMeasurement range: -40 to 260°COne year warrantyLength: 40 inches (1 meter)
Catalog No. NC2883575