Learn More
INTERWORLD HIGHWAY LLC Fluke 80PK-22 SureGrip Immersion Temperature Probe
SDP
Supplier:  INTERWORLD HIGHWAY LLC 2098684
Encompass_Preferred
Fluke 80PK-22 SureGrip Immersion Temperature Probe, Type-K Thermocouple, -40 to 1090 Degree C, 8.375 in probe lengthType-K thermocouple for use in liquids and gelsMeasurement range:-40 to 1090°COne year warranty
Catalog No. NC2883578