Learn More
Cole-Parmer Dry-Well Calibrator, -13 to 284F/Insert A
Supplier:  Cole-Parmer 1610151
Encompass
Fluke Calibration 9103-A Dry-Well Calibrator, -13 to 284F/Insert A, These new units beat every other comparable dry-well in the industry in performance, size, weight, convenience, ease of calibration, software and price. In addition, the heating and cooling rate of each of these dry-wells is adjustable from the front panel, thermal switches can be checked by these units for actuation testing and multiple-hole inserts are available for a variety of probe sizes.
Catalog No. NC1610446
Add to Cart
Request a Quote