Learn More
Cole-Parmer Fluke Digital Multimeter, True RMS, IP67 Rated, 1000 V
Catalog No. NC3992495 Shop All Cole-Parmer Products
Encompass
This digital multimeter offers True RMS accuracy for troubleshooting electrical problems in demanding environments. It is engineered with an IP67 rating for waterproof and dustproof protection, and built to withstand a 13 ft (4 m) drop, ensuring durability for industrial applications.
  • True-rms AC voltage and current for accurate measurements on non-linear signals.
  • Measures up to 1,000 V AC/DC and 10 A (20 A for 30 sec).
  • Built-in temperature function and accurate VFD measurements in noisy environments.
  • Min/Max/Avg recording, 250 μs peak capture, and 19,999 count display mode.
  • IP67 rated, withstands 13 ft (4 m) drops, and has a -15°C to +55°C operating range.
  • CAT IV 600 V/CAT III 1000 V safety rating and 800 hr battery life.
Catalog No. NC3992495 Supplier Cole-Parmer Supplier No. 2003646
This digital multimeter offers True RMS accuracy for troubleshooting electrical problems in demanding environments. It is engineered with an IP67 rating for waterproof and dustproof protection, and built to withstand a 13 ft (4 m) drop, ensuring durability for industrial applications.
  • True-rms AC voltage and current for accurate measurements on non-linear signals.
  • Measures up to 1,000 V AC/DC and 10 A (20 A for 30 sec).
  • Built-in temperature function and accurate VFD measurements in noisy environments.
  • Min/Max/Avg recording, 250 μs peak capture, and 19,999 count display mode.
  • IP67 rated, withstands 13 ft (4 m) drops, and has a -15°C to +55°C operating range.
  • CAT IV 600 V/CAT III 1000 V safety rating and 800 hr battery life.