Learn More
Medchemexpress LLC Flumatinib-d3 | >98% | 565.61 | 1 MG
SDP
Catalog No. 5000347797
Encompass_Preferred
Flumatinib-d3 is deuterium labeled Flumatinib. Flumatinib (HHGV678) is an orally available, selective inhibitor of Bcr-Abl. Flumatinib inhibits c-Abl, PDGFRβ, and c-Kit with IC50s of 1.2 nM, 307.6 nM, and 665.5 nM, respectively.
  • Deuterium labeled Flumatinib
  • Orally available
  • Selective inhibitor of Bcr-Abl
  • Inhibits c-Abl with IC50 of 1.2 nM
  • Inhibits PDGFRβ with IC50 of 307.6 nM
  • Inhibits c-Kit with IC50 of 665.5 nM
Catalog No. 50-003-47797 Supplier Medchemexpress LLC Supplier No. HY13904S1MG
Flumatinib-d3 is deuterium labeled Flumatinib. Flumatinib (HHGV678) is an orally available, selective inhibitor of Bcr-Abl. Flumatinib inhibits c-Abl, PDGFRβ, and c-Kit with IC50s of 1.2 nM, 307.6 nM, and 665.5 nM, respectively.
  • Deuterium labeled Flumatinib
  • Orally available
  • Selective inhibitor of Bcr-Abl
  • Inhibits c-Abl with IC50 of 1.2 nM
  • Inhibits PDGFRβ with IC50 of 307.6 nM
  • Inhibits c-Kit with IC50 of 665.5 nM