Learn More
Enzo Life Sciences FlumazenIL-(100mg). CAS: 78755-81-4
SDP
Supplier:  Enzo Life Sciences BMLB1000100
Encompass_Preferred
Benzodiazepine antagonist. Reverses all the effects of benzodiazepine agonists and inverse agonists. Alternative name: Ro 15-1788. Purity: ≥98% (Assay). Solubility: Soluble in ethanol (5 mg/ml warm) or DMSO (25 mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-201-0337