Learn More
Medchemexpress LLC HY-B0009 50mg Medchemexpress, Flumazenil CAS:78755-81-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB000950MG
Encompass_Preferred
Medchemexpress, HY-B0009 50mg Flumazenil CAS:78755-81-4 Flumazenil is a competitive GABAA receptor antagonist, used in the treatment of benzodiazepine overdoses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2345