Learn More
Apexbio Technology LLC Flumazenil 78755-81-4 25mg
SDP
Catalog No. 5000095660
Encompass_Preferred
Flumazenil (CAS 78755-81-4) is a centrally active small molecule that functions as a benzodiazepine receptor antagonist It binds to -aminobutyric acid type A (GABA sub A /sub ) receptors without preferential selectivity for subtypes containing 1 2 3 or 5 subunits By competitively inhibiting benzodiazepine binding at these receptors flumazenil modulates GABAergic neurotransmission in the central nervous system This compound is widely utilized in biomedical research to investigate benzodiazepine pharmacology GABA sub A /sub receptor function and the neural mechanisms underlying anxiolytic and sedative drug actions
Catalog No. 50-000-95660 Supplier Apexbio Technology LLC Supplier No. A291725
Flumazenil (CAS 78755-81-4) is a centrally active small molecule that functions as a benzodiazepine receptor antagonist It binds to -aminobutyric acid type A (GABA sub A /sub ) receptors without preferential selectivity for subtypes containing 1 2 3 or 5 subunits By competitively inhibiting benzodiazepine binding at these receptors flumazenil modulates GABAergic neurotransmission in the central nervous system This compound is widely utilized in biomedical research to investigate benzodiazepine pharmacology GABA sub A /sub receptor function and the neural mechanisms underlying anxiolytic and sedative drug actions