Learn More
Medchemexpress LLC Flumazenil | 78755-81-4 | >99.9% | C15H14FN3O3 | 5 MG
SDP
Catalog No. 5000432534
Encompass_Preferred
Flumazenil is a competitive GABAA receptor antagonist, primarily used in the treatment of benzodiazepine overdoses. It works by interacting at the central benzodiazepine receptor to antagonize or reverse the behavioral, neurologic, and electrophysiologic effects caused by benzodiazepine agonists and inverse agonists.
  • Functions as a competitive GABAA receptor antagonist
  • Clinically used to treat benzodiazepine overdoses
  • Reverses behavioral, neurologic, and electrophysiologic effects of benzodiazepine agonists
  • May offer benefit in hepatic encephalopathy, pending further clinical trials
  • Induces a strong anxiolytic effect in certain mice models
  • Antagonizes anticonvulsant and adverse effects of diazepam
  • Blocks changes associated with withdrawal from chronic ethanol treatment
Catalog No. 50-004-32534 Supplier Medchemexpress LLC Supplier No. HYB00095MG
Flumazenil is a competitive GABAA receptor antagonist, primarily used in the treatment of benzodiazepine overdoses. It works by interacting at the central benzodiazepine receptor to antagonize or reverse the behavioral, neurologic, and electrophysiologic effects caused by benzodiazepine agonists and inverse agonists.
  • Functions as a competitive GABAA receptor antagonist
  • Clinically used to treat benzodiazepine overdoses
  • Reverses behavioral, neurologic, and electrophysiologic effects of benzodiazepine agonists
  • May offer benefit in hepatic encephalopathy, pending further clinical trials
  • Induces a strong anxiolytic effect in certain mice models
  • Antagonizes anticonvulsant and adverse effects of diazepam
  • Blocks changes associated with withdrawal from chronic ethanol treatment