Learn More
Medchemexpress LLC Flumazenil | 78755-81-4 | 99.99% | C15H14FN3O3 | 1 ML
SDP
Catalog No. 5000433444
Encompass_Preferred
Flumazenil is a competitive GABAA receptor antagonist used in the treatment of benzodiazepine overdoses. This compound is known for its ability to reverse the effects of benzodiazepine agonists and has been explored for various neurological applications.
  • Acts as a competitive GABAA receptor antagonist.
  • Used in the treatment of benzodiazepine overdoses.
  • Reverses behavioral, neurological, and electrophysiological effects of benzodiazepine agonists.
  • May offer benefits in hepatic encephalopathy.
  • Induces anxiolytic effects.
  • Antagonizes anticonvulsant and adverse effects of diazepam.
Catalog No. 50-004-33444 Supplier Medchemexpress LLC Supplier No. HYB000910MM/1ML
Flumazenil is a competitive GABAA receptor antagonist used in the treatment of benzodiazepine overdoses. This compound is known for its ability to reverse the effects of benzodiazepine agonists and has been explored for various neurological applications.
  • Acts as a competitive GABAA receptor antagonist.
  • Used in the treatment of benzodiazepine overdoses.
  • Reverses behavioral, neurological, and electrophysiological effects of benzodiazepine agonists.
  • May offer benefits in hepatic encephalopathy.
  • Induces anxiolytic effects.
  • Antagonizes anticonvulsant and adverse effects of diazepam.