Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flumazenil | 78755-81-4 | 99.99% | C15H14FN3O3 | 100 MG
SDP

Catalog No. 5000432388
Encompass_Preferred

Flumazenil is a competitive GABAA receptor antagonist primarily used to treat benzodiazepine overdoses. It works by interacting with the central benzodiazepine receptor to reverse the effects of benzodiazepine agonists.

  • Reverses sedation from benzodiazepine overdoses
  • Competitive GABAA receptor antagonist
  • Solid appearance
  • Melting point 201-203°C
  • Soluble in DMSO (20 mg/mL)
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • Solution storage at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-32388 Supplier Medchemexpress LLC Supplier No. HYB0009100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flumazenil is a competitive GABAA receptor antagonist primarily used to treat benzodiazepine overdoses. It works by interacting with the central benzodiazepine receptor to reverse the effects of benzodiazepine agonists.

  • Reverses sedation from benzodiazepine overdoses
  • Competitive GABAA receptor antagonist
  • Solid appearance
  • Melting point 201-203°C
  • Soluble in DMSO (20 mg/mL)
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • Solution storage at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.