Learn More
Medchemexpress LLC Flumazenil acid | 84378-44-9 | MFCD08061420 | 100.0% | 275.24 | C13H10FN3O3 | 100 MG
SDP
Catalog No. 5000228583
Encompass_Preferred
Flumazenil acid is the acidic metabolite of the benzodiazepine-site antagonist flumazenil (Ro 15-3890). Supplied primarily as a solid analytical standard, and optionally as a 10 mM solution in DMSO, it is intended for research and analytical use in biochemical and pharmacology studies.
  • Contains flumazenil acid (CAS 84378-44-9) for identity-specific studies.
  • High purity suitable for analytical reference use.
  • Available in multiple pack sizes including 100 mg solid and solution formats.
  • Useful as an LC-MS, HPLC, or reference standard in metabolite and drug metabolism research.
  • Stable solid form with defined molecular weight for quantitative assays.
Catalog No. 50-002-28583 Supplier Medchemexpress LLC Supplier No. HY118844100MG
Flumazenil acid is the acidic metabolite of the benzodiazepine-site antagonist flumazenil (Ro 15-3890). Supplied primarily as a solid analytical standard, and optionally as a 10 mM solution in DMSO, it is intended for research and analytical use in biochemical and pharmacology studies.
  • Contains flumazenil acid (CAS 84378-44-9) for identity-specific studies.
  • High purity suitable for analytical reference use.
  • Available in multiple pack sizes including 100 mg solid and solution formats.
  • Useful as an LC-MS, HPLC, or reference standard in metabolite and drug metabolism research.
  • Stable solid form with defined molecular weight for quantitative assays.