Learn More
AdipoGen Flumequine
SDP
Catalog No. 502560894
Encompass_Preferred
Chemical. CAS 42835-25-6. Formula C14H12FNO3. MW 261.3. Synthetic. Flumequine is a fluoroquinolone synthetic chemotherapeutic antibiotic used to treat bacterial infections. Targets primarily gram negative bacteria, especially those which cause enteric infections in animals. It is used to study processes that affect mammalian chromosome and DNA unwinding at the level of gyrase/topoisomerases. It inhibits topoisomerases, which are needed for the transcription and replication of bacterial DNA. The inhibition of the topoisomerases results in strand breakage of the bacterial chromosome, supercoiling and resealing. Therefore, DNA replication and transcription is inhibited. It was also used to study hepatocarcinogenicity and DNA damage in mice and the mechanisms of quinolone resistance.
Catalog No. 50-256-0894 Supplier AdipoGen Supplier No. CDXF0079G025
Chemical. CAS 42835-25-6. Formula C14H12FNO3. MW 261.3. Synthetic. Flumequine is a fluoroquinolone synthetic chemotherapeutic antibiotic used to treat bacterial infections. Targets primarily gram negative bacteria, especially those which cause enteric infections in animals. It is used to study processes that affect mammalian chromosome and DNA unwinding at the level of gyrase/topoisomerases. It inhibits topoisomerases, which are needed for the transcription and replication of bacterial DNA. The inhibition of the topoisomerases results in strand breakage of the bacterial chromosome, supercoiling and resealing. Therefore, DNA replication and transcription is inhibited. It was also used to study hepatocarcinogenicity and DNA damage in mice and the mechanisms of quinolone resistance.