Learn More
Medchemexpress LLC Flumequine-13C3 | 1185049-09-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0526S1MG
Encompass_Preferred
Flumequine-13C3 is the 13C3 labeled Flumequine. It is a quinolone antibiotic that acts as a topoisomerase II inhibitor, with an IC50 of 15 μM (3.92 μg/mL).
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Applicable in isotope-labeled compounds research
  • Suitable for metabolic flux analysis
  • Used in therapeutic drug monitoring
Catalog No. 50-004-07961