Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flumequine, MP Biomedicals™

Catalog No. ICN15805601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15805601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15805601 Supplier MP Biomedicals, Inc Supplier No. 0215805601

Chemical Identifiers

CAS 42835-25-6
Molecular Formula C14H12FNO3
Molecular Weight (g/mol) 261.252
InChI Key DPSPPJIUMHPXMA-UHFFFAOYSA-N
Synonym flumequine, flumigal, apurone, 9-fluoro-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido 3,2,1-ij quinoline-2-carboxylic acid, flumequinum, flumequino, flumiquil, flumisol, imequyl, flumix
PubChem CID 3374
ChEBI CHEBI:85269
SMILES CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 261.25
Physical Form Crystalline Powder
Chemical Name or Material Flumequine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.