Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flumequine 98.0+%, TCI America™
SDP

Catalog No. F083225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F083225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F083225G Supplier TCI America Supplier No. F083225G

missing translation for 'validMainframeMsg'

Specifications

CAS 42835-25-6
Melting Point 259°C
Color White
Molecular Formula C14H12FNO3
Quantity 25 g
MDL Number MFCD00079298
Synonym flumequine, flumigal, apurone, 9-fluoro-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido 3,2,1-ij quinoline-2-carboxylic acid, flumequinum, flumequino, flumiquil, flumisol, imequyl, flumix
InChI Key DPSPPJIUMHPXMA-UHFFFAOYSA-N
SMILES CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O
Molecular Weight (g/mol) 261.252
PubChem CID 3374
ChEBI CHEBI:85269
Formula Weight 261.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Flumequine
Show More Show Less
RTECSNumber DK1672000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.