Learn More
Medchemexpress LLC Flumethasone | 2135-17-3 | 99.8% | 250 MG
SDP
Catalog No. 5000414250
Encompass_Preferred
Flumethasone (Flumetasone) is an orally active, highly selective and potent glucocorticoid receptor (GR) agonist. It activates GR to inhibit nuclear factor kappa B (NF-κB)-mediated pro-inflammatory cytokine production (TNF-α, IL-1β) and promotes anti-inflammatory gene expression (IL-10). It also regulates metabolic enzyme activity, such as tyrosine aminotransferase induction. Flumethasone shows promise for research into inflammatory diseases, cancer, and endocrine regulation.
  • Orally active
  • High selective and potent glucocorticoid receptor (GR) agonist
  • Inhibits NF-κB-mediated pro-inflammatory cytokine production (TNF-α, IL-1β)
  • Promotes anti-inflammatory gene expression (IL-10)
  • Regulates metabolic enzyme activity (tyrosine aminotransferase induction)
  • Promising for research of inflammatory diseases, cancer, and endocrine regulation
Catalog No. 50-004-14250 Supplier Medchemexpress LLC Supplier No. 250MGHYB1051
Flumethasone (Flumetasone) is an orally active, highly selective and potent glucocorticoid receptor (GR) agonist. It activates GR to inhibit nuclear factor kappa B (NF-κB)-mediated pro-inflammatory cytokine production (TNF-α, IL-1β) and promotes anti-inflammatory gene expression (IL-10). It also regulates metabolic enzyme activity, such as tyrosine aminotransferase induction. Flumethasone shows promise for research into inflammatory diseases, cancer, and endocrine regulation.
  • Orally active
  • High selective and potent glucocorticoid receptor (GR) agonist
  • Inhibits NF-κB-mediated pro-inflammatory cytokine production (TNF-α, IL-1β)
  • Promotes anti-inflammatory gene expression (IL-10)
  • Regulates metabolic enzyme activity (tyrosine aminotransferase induction)
  • Promising for research of inflammatory diseases, cancer, and endocrine regulation