Learn More
Apexbio Technology LLC Flumethasone, 50mg. Cas: 2135-17-3 MFCD: MFCD00056464. Glucocorticoid receptor agonist
SDP
Supplier:  Apexbio Technology LLC B176150
Encompass_Preferred
Flumethasone is a glucocorticoid receptor agonist, this complex binds to the nucleus causing a variety of genetic activation and repressions. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5283