Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flumetsulam | 98967-40-9 | MFCD04089841 | ≥98.0% | 325.29 g/mol | C12H9F2N5O2S | 250 MG
SDP

Catalog No. 5000209657
Encompass_Preferred

Flumetsulam is the analytical reference standard of the sulfonanilide herbicide flumetsulam, provided for research and analytical applications such as method development, identification, and quantitation. It is used as a calibrated standard for chromatographic and mass-spectrometric analysis of environmental and agricultural samples.

  • Analytical standard grade for research and method development.
  • Assay ≥98.0% (HPLC).
  • CAS number 98967-40-9.
  • Molecular weight 325.29 g/mol.
  • Molecular formula C12H9F2N5O2S.
  • Supplied as a 250 mg single package (supplier row information).
  • Suitable for chromatography and quantitative analysis.

Catalog No. 50-002-09657 Supplier Medchemexpress LLC Supplier No. 250MGHYW040303
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flumetsulam is the analytical reference standard of the sulfonanilide herbicide flumetsulam, provided for research and analytical applications such as method development, identification, and quantitation. It is used as a calibrated standard for chromatographic and mass-spectrometric analysis of environmental and agricultural samples.

  • Analytical standard grade for research and method development.
  • Assay ≥98.0% (HPLC).
  • CAS number 98967-40-9.
  • Molecular weight 325.29 g/mol.
  • Molecular formula C12H9F2N5O2S.
  • Supplied as a 250 mg single package (supplier row information).
  • Suitable for chromatography and quantitative analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.